siRNA Details
Gene Symbol: Plod2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001142916.1 | NP_001136388.1 | 13 | 1681 | ||
NM_011961.3 | NP_036091.2 | 13 | 1681 | |||
GenBank mRNA | AF080572.1 | - | 13 | 1452 | ||
AK033363.1 | - | 13 | 1640 | |||
AK046364.1 | - | 13 | 1685 | |||
AK157207.1 | - | 13 | 1641 | |||
BC021352.1 | - | 13 | 1513 | 5007509 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
