siRNA Details
Gene Symbol: Psma7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001289476.1 | NP_001276405.1 | 3 | 970 | ||
NM_011969.2 | NP_036099.1 | 3 | 331 | |||
GenBank mRNA | AF019662.1 | - | 3 | 269 | ||
AK088615.1 | - | 3 | 276 | |||
AK146765.1 | - | 3 | 288 | |||
AK165563.1 | - | 3 | 284 | |||
AK168234.1 | - | 3 | 302 | |||
BC008222.1 | - | 3 | 264 | 2581897 | ||
BC014752.1 | - | 3 | 918 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
