siRNA Details
Gene Symbol: Psmb2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011970.4 | NP_036100.3 | 1 | 188 | ||
GenBank mRNA | AF060090.1 | - | 1 | 76 | ||
AK078487.1 | - | 1 | 167 | |||
AK164463.1 | - | 1 | 165 | |||
BB853639.1 | - | 1 | 201 | |||
BC008265.1 | - | 1 | 115 | 2811010 | ||
CB321399.1 | - | 1 | 184 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
