siRNA Details
Gene Symbol: Psmb3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011971.4 | NP_036101.1 | 4, 5 | 527 | ||
GenBank mRNA | AF060092.1 | - | 4, 5 | 489 | ||
AK002331.1 | - | 4, 5 | 528 | |||
AK010488.1 | - | 4, 5 | 508 | |||
AK153162.1 | - | 4, 5 | 560 | |||
BC014783.1 | - | 4, 5 | 494 | 3500033 | ||
BC098233.1 | - | 3, 4 | 489 | 1547611 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
