siRNA Details
Gene Symbol: Sema4g | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011976.1 | NP_036106.1 | 5 | 848 | ||
GenBank mRNA | AF134918.1 | - | 5 | 848 | ||
AK039355.1 | - | 6 | 932 | |||
AK173219.1 | - | 5 | 3675 | |||
BC049183.1 | - | 5 | 841 | 6395010 | ||
BC054532.1 | - | 6 | 886 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
