siRNA Details
Gene Symbol: Homer2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001164086.1 | NP_001157558.1 | 9 | 1067 | ||
NM_001164087.1 | NP_001157559.1 | 9 | 1083 | |||
NM_011983.2 | NP_036113.1 | 9 | 1100 | |||
GenBank mRNA | AB017136.1 | - | 9 | 1093 | ||
AF093259.1 | - | 9 | 972 | |||
AF093260.1 | - | 9 | 1005 | |||
AK003632.1 | - | 1 | 13 | |||
AK158673.1 | - | 1169 | ||||
BC038314.1 | - | 5 | 741 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
