siRNA Details
Gene Symbol: Ncdn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011986.4 | NP_036116.3 | 3 | 877 | ||
GenBank mRNA | AK122327.1 | - | 1 | 222 | ||
AK154889.1 | - | 3 | 858 | |||
AK170502.1 | - | 4 | 592 | |||
BC017126.1 | - | 3 | 808 | 4485839 | ||
BQ745150.1 | - | 2 | 307 | |||
CX568560.1 | - | 1 | 263 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
