siRNA Details
Gene Symbol: Cenph | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021886.1 | NP_068686.1 | 3 | 306 | ||
GenBank mRNA | AB017634.1 | - | 3 | 306 | ||
AK006208.1 | - | 3 | 290 | |||
AK011264.1 | - | 3 | 291 | |||
AK011738.1 | - | 3 | 291 | |||
BC025084.1 | - | 3 | 257 | 3501244 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
