siRNA Details
Gene Symbol: Cln8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012000.3 | NP_036130.1 | 3 | 1426 | ||
GenBank mRNA | AF125307.1 | - | 3 | 916 | ||
AF125308.1 | - | 3 | 916 | |||
AK049145.1 | - | 3 | 1099 | |||
AK080049.1 | - | 3 | 934 | |||
AK088300.1 | - | 3 | 1080 | |||
AK089692.1 | - | 3 | 1075 | |||
AK128890.1 | - | 3 | 1097 | |||
AK164268.1 | - | 3 | 1099 | |||
AK166905.1 | - | 3 | 1851 | |||
AK169698.1 | - | 3 | 1080 | |||
AK171446.1 | - | 3 | 1108 | |||
AK201598.1 | - | 1 | 164 | |||
BC021625.1 | - | 3 | 932 | 4188527 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
