siRNA Details
Gene Symbol: Dysf | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001077694.2 | NP_001071162.1 | 26 | 3108 | ||
NM_021469.3 | NP_067444.2 | 26 | 2926 | |||
GenBank mRNA | AF188290.2 | - | 25 | 2701 | ||
AK034587.1 | - | 5 | 719 | |||
AK131144.1 | - | 26 | 2923 |
