siRNA Details
Gene Symbol: Gcat | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001161712.1 | NP_001155184.1 | 2, 3 | 383 | ||
NM_013847.4 | NP_038875.3 | 2, 3 | 383 | |||
GenBank mRNA | AF093403.1 | - | 2, 3 | 341 | ||
AK013138.1 | - | 2, 3 | 338 | |||
AK075782.1 | - | 2, 3 | 323 | |||
AK076469.1 | - | 2, 3 | 356 | |||
AK087433.1 | - | 2, 3 | 342 | |||
AK183376.1 | - | 1, 2 | 158 | |||
AK187910.1 | - | 2, 3 | 159 | |||
AK219228.1 | - | 2, 3 | 158 | |||
BC024107.1 | - | 2, 3 | 383 | 5125361 |

More Information
Other Species Targeted

Gene Ontology Categories:
