siRNA Details
Gene Symbol: Cntrl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001290635.1 | NP_001277564.1 | 4 | 711 | ||
NM_012018.2 | NP_036148.2 | 14 | 2353 | |||
GenBank mRNA | AK088733.1 | - | 4 | 713 | ||
BC050831.1 | - | 6 | 985 | |||
BC060146.1 | - | 4 | 687 | |||
BC079536.1 | - | 2 | 375 | |||
BC158010.1 | - | 13 | 2226 | 9007318 | ||
X79131.1 | - | 6 | 1013 |
