siRNA Details
Gene Symbol: Map4k4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001252200.1 | NP_001239129.1 | 4 | 550 | ||
NM_001252201.1 | NP_001239130.1 | 4 | 550 | |||
NM_001252202.1 | NP_001239131.1 | 4 | 550 | |||
NM_008696.2 | NP_032722.2 | 4 | 550 | |||
GenBank mRNA | AK129191.1 | - | 4 | 287 | ||
BC056650.1 | - | 4 | 293 | |||
BC141109.1 | - | 4 | 284 | 9055721 | ||
BC145390.1 | - | 4 | 284 | 9053948 | ||
BC145391.1 | - | 4 | 283 | 9053949 | ||
CF531764.1 | - | 2 | 108 | |||
CJ142598.1 | - | 4 | 557 | |||
U88984.1 | - | 4 | 230 |
