siRNA Details
Gene Symbol: Spag1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012031.3 | NP_036161.2 | 18 | 2570 | ||
GenBank mRNA | AF181252.1 | - | 18 | 2562 | ||
AF181253.1 | - | 8 | 1254 | |||
AK032601.1 | - | 15 | 2365 | |||
BC046313.1 | - | 18 | 2570 | 6310604 | ||
BC098201.1 | - | 2 | 1900 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
