siRNA Details
Gene Symbol: Spag1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012031.3 | NP_036161.2 | 12 | 1658 | ||
GenBank mRNA | AF181252.1 | - | 12 | 1650 | ||
AF181253.1 | - | 2 | 342 | |||
AK032601.1 | - | 9 | 1453 | |||
BC046313.1 | - | 12 | 1658 | 6310604 |

More Information
Other Species Targeted

Gene Ontology Categories:
