siRNA Details
Gene Symbol: Serinc3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012032.4 | NP_036162.3 | 5 | 677 | ||
GenBank mRNA | AB029499.1 | - | 5 | 586 | ||
AF181684.1 | - | 5 | 605 | |||
AK031101.1 | - | 5 | 666 | |||
AK128955.1 | - | 3 | 214 | |||
AK145604.1 | - | 5 | 625 | |||
AK150106.1 | - | 5 | 623 | |||
AK152720.1 | - | 5 | 623 | |||
AK155921.1 | - | 5 | 623 | |||
AK159347.1 | - | 5 | 623 | |||
AK159488.1 | - | 5 | 623 | |||
AK159819.1 | - | 5 | 623 | |||
AK160005.1 | - | 5 | 623 | |||
AK160055.1 | - | 5 | 623 | |||
AK161981.1 | - | 5 | 623 | |||
AK164807.1 | - | 5 | 623 | |||
AK166924.1 | - | 5 | 623 | |||
AK170350.1 | - | 5 | 616 | |||
BC011295.1 | - | 5 | 633 | 3487943 | ||
BC022901.1 | - | 5 | 614 | 5068196 | ||
BC029026.1 | - | 5 | 642 | 5039323 | ||
BC029980.1 | - | 5 | 646 | |||
BC058550.1 | - | 350 | ||||
L29441.1 | - | 4 | 402 |

More Information
Other Species Targeted

Gene Ontology Categories:
