siRNA Details
Gene Symbol: Cul1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012042.3 | NP_036172.1 | 14 | 1842 | ||
GenBank mRNA | AF136441.1 | - | 13 | 1697 | ||
AF176910.1 | - | 14 | 1731 | |||
AK147004.1 | - | 14 | 1824 | |||
AK164274.1 | - | 14 | 1809 | |||
AK165978.1 | - | 14 | 1783 | |||
BC004836.1 | - | 13 | 1651 | |||
BC029260.1 | - | 14 | 1842 | 2812400 |
