siRNA Details
Gene Symbol: Brd7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012047.2 | NP_036177.1 | 6 | 884 | ||
GenBank mRNA | AF084259.1 | - | 6 | 857 | ||
AK138934.1 | - | 6 | 888 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
