siRNA Details
Gene Symbol: Tspan32 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001128080.2 | NP_001121552.1 | 7, 8 | 698 | ||
NM_020286.3 | NP_064682.1 | 6, 7 | 813 | |||
GenBank mRNA | AF175771.1 | - | 6, 7 | 662 | ||
AF291425.1 | - | 6, 7 | 781 | |||
AJ279791.1 | - | 6, 7 | 773 | |||
AK154694.1 | - | 6, 7 | 834 | |||
AY039000.1 | - | 7, 8 | 706 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
