siRNA Details
Gene Symbol: Tspan32 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001128080.2 | NP_001121552.1 | 8 | 754 | ||
NM_001128081.1 | NP_001121553.1 | 6 | 785 | |||
NM_001128082.1 | NP_001121554.1 | 5 | 683 | |||
NM_020286.3 | NP_064682.1 | 7 | 869 | |||
GenBank mRNA | AF175771.1 | - | 7 | 718 | ||
AF291425.1 | - | 7 | 837 | |||
AJ279791.1 | - | 7 | 829 | |||
AJ279792.1 | - | 6 | 745 | |||
AJ279794.1 | - | 5 | 643 | |||
AK013248.1 | - | 6 | 560 | |||
AK154694.1 | - | 7 | 890 | |||
AK171199.1 | - | 7 | 974 | |||
AY039000.1 | - | 8 | 762 | |||
BC051204.1 | - | 5 | 673 | |||
BC055858.1 | - | 6 | 759 | 4913671 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
