siRNA Details
Gene Symbol: Tspan32 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001128080.2 | NP_001121552.1 | 5 | 456 | ||
NM_001128081.1 | NP_001121553.1 | 4 | 571 | |||
NM_020286.3 | NP_064682.1 | 4 | 571 | |||
GenBank mRNA | AF175771.1 | - | 4 | 420 | ||
AF291425.1 | - | 4 | 539 | |||
AJ279791.1 | - | 4 | 531 | |||
AJ279792.1 | - | 4 | 531 | |||
AJ279793.1 | - | 4 | 531 | |||
AK013248.1 | - | 4 | 347 | |||
AK154694.1 | - | 4 | 592 | |||
AK171199.1 | - | 5 | 760 | |||
AY039000.1 | - | 5 | 464 | |||
BC055858.1 | - | 4 | 545 | 4913671 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
