siRNA Details
Gene Symbol: G3bp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013716.2 | NP_038744.1 | 9 | 980 | ||
GenBank mRNA | AB001927.1 | - | 9 | 980 | ||
AK141689.1 | - | 9 | 1047 | |||
AK182641.1 | - | 2 | 176 | |||
AK220230.1 | - | 9 | 1034 | |||
BC021156.1 | - | 9 | 992 | 4020362 |
