siRNA Details
Gene Symbol: Etv3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001083318.2 | NP_001076787.1 | 4 | 423 | ||
NM_001286844.1 | NP_001273773.1 | 4 | 739 | |||
NM_012051.4 | NP_036181.3 | 4 | 420 | |||
GenBank mRNA | AF156530.1 | - | 3 | 349 | ||
AF218539.2 | - | 4 | 406 | |||
AK089640.1 | - | 4 | 370 | |||
AK147552.1 | - | 2 | 2546 | |||
AK155080.1 | - | 2 | 2053 | |||
AK155957.1 | - | 4 | 361 | |||
AK156824.1 | - | 4 | 368 | |||
AK163509.1 | - | 4 | 388 | |||
BC057151.1 | - | 4 | 357 | |||
BC078636.1 | - | 4 | 357 | 4918332 |

More Information
Other Species Targeted

Gene Ontology Categories:
