siRNA Details
Gene Symbol: Fkbp9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012056.2 | NP_036186.2 | 8 | 1386 | ||
GenBank mRNA | AF090334.1 | - | 8 | 1312 | ||
AF279263.1 | - | 8 | 1364 | |||
AK007499.1 | - | 6 | 776 | |||
BC026133.1 | - | 8 | 1326 | 4018235 | ||
BC043129.1 | - | 8 | 1386 | 6810029 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
