siRNA Details
Gene Symbol: Srp9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012058.3 | NP_036188.1 | 2 | 291 | ||
GenBank mRNA | AK011720.1 | - | 2 | 219 | ||
AK020583.1 | - | 2 | 240 | |||
AK020620.1 | - | 2 | 232 | |||
AK051208.1 | - | 2 | 237 | |||
AK146882.1 | - | 2 | 240 | |||
AK169314.1 | - | 2 | 239 | |||
AK187269.1 | - | 1 | 66 | |||
BC039648.1 | - | 2 | 278 | 5353755 | ||
BC090254.1 | - | 2 | 219 | 5100716 | ||
X78304.1 | - | 2 | 217 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
