siRNA Details
Gene Symbol: Sh3d19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001082414.2 | NP_001075883.2 | 11 | 2021 | ||
GenBank mRNA | AF003234.1 | - | 5 | 411 | ||
AK050185.1 | - | 3 | 278 | |||
BC028254.1 | - | 11 | 1569 | |||
BC031117.1 | - | 11 | 1132 | 5064197 | ||
D89677.2 | - | 17 | 2507 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
