siRNA Details
Gene Symbol: Eif2ak4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001177806.1 | NP_001171277.1 | 32 | 4233 | ||
NM_013719.3 | NP_038747.2 | 33 | 4537 | |||
GenBank mRNA | AJ243533.1 | - | 33 | 4537 | ||
AK011380.1 | - | 6 | 623 | |||
AK129334.1 | - | 28 | 3887 | |||
BC072637.1 | - | 32 | 4233 | 30354433 | ||
W75391.1 | - | 3 | 239 |
