siRNA Details
Gene Symbol: Podxl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013723.3 | NP_038751.2 | 5, 6 | 1367 | ||
GenBank mRNA | AB028048.1 | - | 5, 6 | 1248 | ||
AF290209.1 | - | 5, 6 | 1266 | |||
AK134723.1 | - | 1, 2 | 103 | |||
BC052442.1 | - | 5, 6 | 1341 | 6406610 | ||
BC054530.1 | - | 5, 6 | 1341 | 6406610 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
