siRNA Details
Gene Symbol: Cartpt | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081493.2 | NP_001074962.1 | 2 | 251 | ||
NM_013732.7 | NP_038760.3 | 2 | 290 | |||
GenBank mRNA | AK134656.1 | - | 2 | 226 | ||
AK135007.1 | - | 2 | 223 | |||
BC056431.1 | - | 2 | 248 | 6817832 | ||
CJ172339.1 | - | 2 | 252 | |||
CO427341.1 | - | 2 | 619 |

More Information
Other Species Targeted

Gene Ontology Categories:
