siRNA Details
Gene Symbol: Cartpt | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081493.2 | NP_001074962.1 | 3 | 765 | ||
NM_013732.7 | NP_038760.3 | 3 | 804 | |||
GenBank mRNA | AK134656.1 | - | 3 | 740 | ||
AV341177.1 | - | 1 | 185 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
