siRNA Details
Gene Symbol: Chaf1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013733.3 | NP_038761.1 | 3 | 277 | ||
GenBank mRNA | AJ132771.2 | - | 3 | 181 | ||
AK034839.1 | - | 3 | 278 | |||
AK132551.1 | - | 101 | ||||
AK135763.1 | - | 3 | 290 | |||
BC053740.1 | - | 3 | 248 | 6504838 |
