siRNA Details
Gene Symbol: Tceb3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013736.4 | NP_038764.2 | 9 | 2094 | ||
GenBank mRNA | AB025015.1 | - | 9 | 2078 | ||
AK036592.1 | - | 9 | 2095 | |||
AK145994.1 | - | 9 | 2095 | |||
AK147088.1 | - | 9 | 2089 | |||
BC049885.1 | - | 9 | 2090 | 5387774 |
