siRNA Details
Gene Symbol: Cars | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001252593.1 | NP_001239522.1 | 16, 17 | 1816 | ||
NM_013742.5 | NP_038770.3 | 17, 18 | 2065 | |||
GenBank mRNA | AJ276796.1 | - | 17, 18 | 2032 | ||
AK033328.1 | - | 17, 18 | 2032 | |||
AK037227.1 | - | 16, 17 | 1784 | |||
AK047027.1 | - | 16, 17 | 1784 | |||
AK077558.1 | - | 16, 17 | 1782 | |||
AK133205.1 | - | 16, 17 | 1783 | |||
AK135433.1 | - | 17, 18 | 2035 | |||
AK152866.1 | - | 16, 17 | 1789 | |||
BC030473.1 | - | 13, 14 | 1501 | |||
BC054717.1 | - | 17, 18 | 2014 | 5686364 | ||
BC056194.1 | - | 3, 4 | 367 | |||
BC058954.1 | - | 17, 18 | 2017 | 5707198 |
