siRNA Details
Gene Symbol: Cars | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001252593.1 | NP_001239522.1 | 17 | 1859 | ||
NM_013742.5 | NP_038770.3 | 18 | 2108 | |||
GenBank mRNA | AJ276796.1 | - | 18 | 2075 | ||
AK033328.1 | - | 18 | 2075 | |||
AK037227.1 | - | 17 | 1827 | |||
AK047027.1 | - | 17 | 1827 | |||
AK077558.1 | - | 17 | 1825 | |||
AK133205.1 | - | 17 | 1826 | |||
AK135433.1 | - | 18 | 2078 | |||
AK152866.1 | - | 17 | 1832 | |||
BC030473.1 | - | 14 | 1544 | |||
BC054717.1 | - | 18 | 2057 | 5686364 | ||
BC056194.1 | - | 4 | 410 | |||
BC058954.1 | - | 18 | 2060 | 5707198 |
