siRNA Details
Gene Symbol: Cars | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001252593.1 | NP_001239522.1 | 14 | 1551 | ||
NM_013742.5 | NP_038770.3 | 15 | 1800 | |||
GenBank mRNA | AB015589.1 | - | 12 | 1334 | ||
AJ276796.1 | - | 15 | 1767 | |||
AK033328.1 | - | 15 | 1767 | |||
AK037227.1 | - | 14 | 1519 | |||
AK047027.1 | - | 14 | 1519 | |||
AK077558.1 | - | 14 | 1517 | |||
AK133205.1 | - | 14 | 1518 | |||
AK135433.1 | - | 15 | 1770 | |||
AK152866.1 | - | 14 | 1524 | |||
AK198552.1 | - | 4 | 362 | |||
BC030473.1 | - | 11 | 1236 | |||
BC054717.1 | - | 15 | 1749 | 5686364 | ||
BC056194.1 | - | 1 | 102 | |||
BC058954.1 | - | 15 | 1752 | 5707198 |
