siRNA Details
Gene Symbol: Zfp354b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013744.3 | NP_038772.1 | 5 | 1931 | ||
GenBank mRNA | AF184112.1 | - | 4 | 1800 | ||
AK010007.1 | - | 1 | 1018 | |||
AK148756.1 | - | 5 | 1934 | |||
BC107400.1 | - | 4 | 1768 | 40058571 | ||
BC107401.1 | - | 4 | 1768 | 40058574 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
