siRNA Details
Gene Symbol: Nbn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013752.3 | NP_038780.3 | 13 | 2088 | ||
GenBank mRNA | AB016988.1 | - | 13 | 2010 | ||
AF076687.1 | - | 13 | 2082 | |||
AF092840.1 | - | 13 | 2071 | |||
AK132302.1 | - | 13 | 2130 | |||
AK159790.1 | - | 13 | 2094 | |||
AK160614.1 | - | 13 | 2055 | |||
BC003719.1 | - | 13 | 2097 | |||
BC055061.1 | - | 13 | 2080 | 6528890 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
