siRNA Details
Gene Symbol: Nbn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013752.3 | NP_038780.3 | 7, 8 | 1011 | ||
GenBank mRNA | AB016988.1 | - | 7, 8 | 933 | ||
AF076687.1 | - | 7, 8 | 1005 | |||
AF092840.1 | - | 7, 8 | 994 | |||
AK031933.1 | - | 7, 8 | 997 | |||
AK132302.1 | - | 7, 8 | 1052 | |||
AK134805.1 | - | 7, 8 | 1047 | |||
AK134960.1 | - | 7, 8 | 1039 | |||
AK159790.1 | - | 7, 8 | 1018 | |||
AK160614.1 | - | 7, 8 | 978 | |||
AK183777.1 | - | 2, 3 | 243 | |||
BC003719.1 | - | 7, 8 | 1020 | |||
BC013190.1 | - | 7, 8 | 1011 | |||
BC044773.2 | - | 7, 8 | 1010 | 5067851 | ||
BC055061.1 | - | 7, 8 | 1003 | 6528890 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
