siRNA Details
Gene Symbol: Pald1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013753.2 | NP_038781.2 | 11 | 1571 | ||
GenBank mRNA | AK088577.1 | - | 11 | 1407 | ||
AK146489.1 | - | 11 | 1516 | |||
BC137695.1 | - | 10 | 1335 | 8860714 | ||
BC144846.1 | - | 10 | 1344 | 9053394 | ||
BC144847.1 | - | 10 | 1341 | 9053395 | ||
X99384.1 | - | 10 | 1356 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
