siRNA Details
Gene Symbol: Msrb1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013759.2 | NP_038787.1 | 2 | 251 | ||
GenBank mRNA | AF195142.1 | - | 2 | 226 | ||
AK002339.1 | - | 2 | 211 | |||
AK002652.1 | - | 2 | 210 | |||
AK003112.1 | - | 2 | 211 | |||
AK003624.1 | - | 2 | 211 | |||
BC090646.1 | - | 2 | 251 | 30946285 | ||
BC141146.1 | - | 2 | 215 | 9055758 | ||
CX566836.1 | - | 2 | 251 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
