siRNA Details
Gene Symbol: Rpl3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013762.2 | NP_038790.2 | 6, 7 | 924 | ||
GenBank mRNA | AK088993.1 | - | 6, 7 | 871 | ||
AK144518.1 | - | 6, 7 | 871 | |||
AK144816.1 | - | 6, 7 | 871 | |||
AK146502.1 | - | 6, 7 | 871 | |||
AK149494.1 | - | 6, 7 | 871 | |||
AK151057.1 | - | 6, 7 | 871 | |||
AK151084.1 | - | 6, 7 | 871 | |||
AK151142.1 | - | 6, 7 | 871 | |||
AK151426.1 | - | 6, 7 | 871 | |||
AK151596.1 | - | 6, 7 | 871 | |||
AK151740.1 | - | 6, 7 | 871 | |||
AK166433.1 | - | 6, 7 | 867 | |||
AK166825.1 | - | 6, 7 | 871 | |||
AK167787.1 | - | 6, 7 | 871 | |||
AK167947.1 | - | 6, 7 | 871 | |||
AK168040.1 | - | 6, 7 | 871 | |||
AK168059.1 | - | 6, 7 | 870 | |||
AK168126.1 | - | 6, 7 | 871 | |||
AK168128.1 | - | 6, 7 | 871 | |||
AK168224.1 | - | 6, 7 | 871 | |||
AK168744.1 | - | 6, 7 | 869 | |||
AK169060.1 | - | 6, 7 | 871 | |||
AK169235.1 | - | 6, 7 | 871 | |||
AK169321.1 | - | 6, 7 | 871 | |||
AK170819.1 | - | 6, 7 | 871 | |||
AK172280.1 | - | 6, 7 | 871 | |||
AK190439.1 | - | 2, 3 | 247 | |||
AK192125.1 | - | 2, 3 | 182 | |||
AK197531.1 | - | 2, 3 | 247 | |||
AK199153.1 | - | 1, 2 | 160 | |||
AK199385.1 | - | 1, 2 | 158 | |||
AK204620.1 | - | 1, 2 | 173 | |||
AK204785.1 | - | 1, 2 | 166 | |||
AK215968.1 | - | 2, 3 | 247 | |||
BC009655.1 | - | 6, 7 | 867 | 3485069 | ||
BC083134.1 | - | 5, 6 | 853 | 6504713 | ||
BC085495.1 | - | 5, 6 | 1375 | |||
BC094059.1 | - | 5, 6 | 852 | 3676103 | ||
BC098497.1 | - | 4, 5 | 1429 | |||
BC158039.1 | - | 6, 7 | 871 | 9007342 | ||
U89417.1 | - | 2, 3 | 299 | |||
Y00225.1 | - | 5, 6 | 853 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | AK219709.1 | - | 359 | Chr. - - [-] on Build |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
