siRNA Details
Gene Symbol: Dguok | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001162521.1 | NP_001155993.1 | 5 | 770 | ||
NM_013764.2 | NP_038792.2 | 5 | 770 | |||
GenBank mRNA | AJ133749.1 | - | 5 | 695 | ||
AK035281.1 | - | 5 | 716 | |||
AK167071.1 | - | 5 | 592 | |||
AY037862.1 | - | 5 | 700 | |||
BC012647.1 | - | 5 | 715 | |||
BC094920.1 | - | 5 | 770 | 30055743 | ||
BQ746075.1 | - | 5 | 684 | |||
U90524.1 | - | 5 | 690 |

More Information
Other Species Targeted

Gene Ontology Categories:
