siRNA Details
Gene Symbol: Tcl1b3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013772.2 | NP_038800.1 | 1 | 180 | ||
GenBank mRNA | AF195490.1 | - | 1 | 182 | ||
BC119260.1 | - | 1 | 180 | 8734009 | ||
BC120575.1 | - | 1 | 180 | 40129498 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
