siRNA Details
Gene Symbol: Tcl1b2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013775.1 | NP_038803.1 | 1 | 173 | ||
GenBank mRNA | AF195489.1 | - | 1 | 173 | ||
AK139548.1 | - | 1 | 188 | |||
BC099461.1 | - | 1 | 157 | 30935073 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
