siRNA Details
Gene Symbol: Tcl1b5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013776.1 | NP_038804.1 | 1 | 154 | ||
GenBank mRNA | AF195493.1 | - | 1 | 154 | ||
AK139627.1 | - | 1 | 169 | |||
BC125294.1 | - | 1 | 151 | 40129809 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
