siRNA Details
Gene Symbol: Abcf2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001190443.1 | NP_001177372.1 | 6 | 1056 | ||
NM_013853.2 | NP_038881.1 | 6 | 1059 | |||
GenBank mRNA | AK087990.1 | - | 6 | 961 | ||
AK137233.1 | - | 6 | 991 | |||
AK147937.1 | - | 6 | 1014 | |||
AK151545.1 | - | 6 | 1015 | |||
BC003300.1 | - | 5 | 951 | 3583837 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
