siRNA Details
Gene Symbol: Slc14a2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_030683.3 | NP_109608.1 | 5 | 670 | ||
NM_207651.3 | NP_997534.2 | 5 | 670 | |||
GenBank mRNA | AF258602.1 | - | 5 | 571 | ||
AF366052.2 | - | 5 | 662 | |||
BC150680.1 | - | 5 | 578 | 9087591 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
