siRNA Details
Gene Symbol: Slc14a2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001110274.1 | NP_001103744.1 | 4 | 587 | ||
NM_030683.3 | NP_109608.1 | 8 | 1213 | |||
NM_207651.3 | NP_997534.2 | 8 | 1213 | |||
GenBank mRNA | AF258601.2 | - | 4 | 587 | ||
AF258602.1 | - | 8 | 1114 | |||
AF366052.2 | - | 8 | 1205 | |||
AY221737.1 | - | 3, 4 | 299 | |||
BC150680.1 | - | 8 | 1121 | 9087591 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
