siRNA Details
Gene Symbol: Peg12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013788.2 | NP_038816.1 | 1 | 1470 | ||
GenBank mRNA | AF148857.1 | - | 1 | 1470 | ||
AK142886.1 | - | 1 | 253 | |||
BC138242.1 | - | 1 | 1434 | 8861261 | ||
BC138243.1 | - | 1 | 1437 | 8861262 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
