siRNA Details
Gene Symbol: Abcc5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013790.2 | NP_038818.2 | 3 | 426 | ||
NM_176839.1 | NP_789809.1 | 3 | 426 | |||
GenBank mRNA | AB019003.1 | - | 3 | 377 | ||
AK050565.1 | - | 3 | 419 | |||
BC042581.1 | - | 3 | 368 | |||
BC061132.1 | - | 3 | 386 | |||
BC090629.1 | - | 3 | 383 | 6839816 | ||
CX240998.1 | - | 3 | 402 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
